3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

D5341-1G D5341-200MG
TCD5341-1GEA 432.92 CAD
TCD5341-1G TCD5341-200MG
3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole ≥98.0% (by total nitrogen basis)
3,6-Dibromo-9-(4-n-octyloxyphenyl)-9H-carbazole
Formula: C₂₆H₂₇Br₂NO
MDL Number: MFCD30536723
CAS Number: 917773-26-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR