Cesium(I) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3264-1G C3264-5G
TCC3264-1GEA 169.06 CAD
TCC3264-1G TCC3264-5G
Cesium(I) bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Cesium(I) bis(trifluoromethanesulfonyl)imide
Formula: C₂CsF₆NO₄S₂
Melting Pt: 122°C
MDL Number: MFCD29472520
CAS Number: 91742-16-4
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR