trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
D4652-1G
D4652-200MG
TCD4652-1GEA
900.92
CAD
TCD4652-1G
TCD4652-200MG
trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline ≥95.0% (by GC)
trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline
Formula:
C₁₈H₂₂N₂ |
MDL Number:
MFCD06669887 CAS Number: 908294-68-8 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...