4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4'-[(6-Acryloyloxy)hexyloxy]-4-biphenylcarbonitrile, 6-[(4'-Cyanobiphenyl-4-yl)oxy]hexyl acrylate, Acrylic acid 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl ester, 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl acrylate, 6-((4'-Cyano-[1,1'-biphenyl]-4-yl)oxy)hexyl acrylate

A2986-1G A2986-5G
TCA2986-1GEA 255.96 CAD
TCA2986-1G TCA2986-5G
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98.0% (by GC)
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl
Formula: C₂₂H₂₃NO₃
MW: 349.43 g/mol
Melting Pt: 71°C
MDL Number: MFCD16293619
CAS Number: 89823-23-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR