N-[4-(1-Naphthyl)phenyl]-4-biphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N-[4-(Naphthalen-1-yl)phenyl]-[1,1'-biphenyl]-4-amine, N-(4-(Naphthalen-1-yl)phenyl)-[1,1'-biphenyl]-4-amine

N1053-1G N1053-200MG
TCN1053-1GEA 641.48 CAD
TCN1053-1G TCN1053-200MG
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine ≥98.0% (by GC)
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine
Formula: C₂₈H₂₁N
MW: 371.48 g/mol
MDL Number: MFCD28138088
CAS Number: 897921-59-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR