[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane) ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2609-1G
M2609-200MG
76220-238EA
783.68
CAD
76220-238
76220-240
[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane) ≥98.0% (by GC)
[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane)
Formula:
C₁₀H₉BF₂N₂S |
CAS Number:
892505-41-8 |
Specification Test Results
Appearance | Red to Dark red to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Lambda max. | 486.0 to 490.0 nm (EtOH) |
Molar extinction coefficient | min. 33000 (EtOH, 486.0 to 490.0nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...