4-Styryltriphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

S0924-1G S0924-5G
TCS0924-1GEA 264.49 CAD
TCS0924-1G TCS0924-5G
4-Styryltriphenylamine ≥98.0% (by GC)
4-Styryltriphenylamine
Formula: C₂₆H₂₁N
MDL Number: MFCD16619204
CAS Number: 89114-74-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR