2,4,6-Tris(3-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

T3308-1G T3308-5G
TCT3308-1GEA 120.08 CAD
TCT3308-1G TCT3308-5G
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine
Formula: C₂₁H₁₂Br₃N₃
MDL Number: MFCD28964709
CAS Number: 890148-78-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 207.0 to 211.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR