4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Triphenylen-2-ylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)triphenylene

T3322-1G T3322-5G
TCT3322-1GEA 264.49 CAD
TCT3322-1G TCT3322-5G
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane ≥97.0% (by HPLC, total nitrogen)
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane
Formula: C₂₄H₂₃BO₂
MW: 354.26 g/mol
MDL Number: MFCD26401844
CAS Number: 890042-13-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 174.0 to 178.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR