2,7-Dibromotriphenylene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4801-1G D4801-200MG
TCD4801-1GEA 272.71 CAD
TCD4801-1G TCD4801-200MG
2,7-Dibromotriphenylene ≥98.0% (by GC)
2,7-Dibromotriphenylene
Formula: C₁₈H₁₀Br₂
MDL Number: MFCD22571695
CAS Number: 888041-37-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 230.0 to 234.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR