3-Aminotetrahydrofuran ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Tetrahydrofuran-3-amine

A2700-1G A2700-250MG
TCA2700-1GEA 377.62 CAD
TCA2700-1G TCA2700-250MG
3-Aminotetrahydrofuran ≥97.0% (by GC)
3-Aminotetrahydrofuran
Formula: C₄H₉NO
Boiling Pt: 73°C
Flash Pt: 49°C
MDL Number: MFCD07778394
CAS Number: 88675-24-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR