2-Phenyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

P2375-1G P2375-5G
TCP2375-1GEA 58.46 CAD
TCP2375-1G TCP2375-5G
2-Phenyl-9H-carbazole ≥98.0% (by GC)
2-Phenyl-9H-carbazole
Formula: C₁₈H₁₃N
MDL Number: MFCD26402145
CAS Number: 88590-00-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR