Ethylhexyl triazone ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Tris(2-ethylhexyl) 4,4',4''-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoate, Tris(2-ethylhexyl) 4,4',4''-((1,3,5-triazine-2,4,6-triyl)tris(azanediyl))tribenzoate, 4,4',4''-(1,3,5-Triazine-2,4,6-triyltriimino)trisbenzoic acid tris(2-ethylhexyl) ester

E1312-1G E1312-5G
TCE1312-1GEA 28.44 CAD
TCE1312-1G TCE1312-5G
Ethylhexyl triazone ≥98.0% (by total nitrogen basis)
Ethylhexyl triazone
Formula: C₄₈H₆₆N₆O₆
MW: 823.09 g/mol
MDL Number: MFCD09753106
CAS Number: 88122-99-0
EINECS: 402-070-1
Merck Index: 14,03809

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 124.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR