1,2,3-Trimethyl-3H-benzo[e]indole ≥98.0% (by GC)
Supplier: TCI America
T3119-25G
T3119-5G
TCT3119-25GEA
267.81
CAD
TCT3119-25G
TCT3119-5G
1,2,3-Trimethyl-3H-benzo[e]indole ≥98.0% (by GC)
Formula:
C₁₅H₁₅N |
MDL Number:
MFCD06651515 CAS Number: 881219-73-4 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...