4-Methylumbelliferyl beta-D-Glucuronide hydrate ≥98.0% (by HPLC)

Supplier: TCI America

M3026-100MG M3026-1G
TCM3026-100MGEA 90.06 CAD
TCM3026-100MG TCM3026-1G
4-Methylumbelliferyl beta-D-Glucuronide hydrate ≥98.0% (by HPLC)
4-Methylumbelliferyl beta-D-Glucuronide hydrate
Formula: C₁₆H₁₆O₉·xH₂O
MDL Number: MFCD09039280
CAS Number: 881005-91-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -115 to -120 deg (C=0.2, H2O) (calcd.on anh.substance)
Water 5.0 to 17.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR