Tris(1H-benzotriazol-1-yl)methane ≥95.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Tris(1H-benzo[d][1,2,3]triazol-1-yl)methane

T3141-1G T3141-200MG
TCT3141-1GEA 388.36 CAD
TCT3141-1G TCT3141-200MG
Tris(1H-benzotriazol-1-yl)methane ≥95.0% (by HPLC)
Tris(1H-benzotriazol-1-yl)methane
Formula: C₁₉H₁₃N₉
MW: 367.38 g/mol
Melting Pt: 191°C
MDL Number: MFCD00514776
CAS Number: 88088-95-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR