N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5247-1G B5247-200MG
TCB5247-1GEA 140.62 CAD
TCB5247-1G TCB5247-200MG
N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline methyl ester
Formula: C₁₈H₂₅NO₇S
Storage Temperature: Refrigerator
MDL Number: MFCD02179383
CAS Number: 88043-21-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 77.0 to 81.0 deg-C
Specific rotation [a]20/D -33.0 to -38.0 deg (C=0.7, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR