2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
P2384-1G
P2384-200MG
TCP2384-1GEA
481.9
CAD
TCP2384-1G
TCP2384-200MG
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98.0% (by GC, titration analysis)
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula:
C₁₇H₂₀BNO₂ |
MDL Number:
MFCD12032565 CAS Number: 879291-27-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...