(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5682-1G B5682-5G
76218-880EA 150.1 CAD
76218-880 76218-882
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine ≥98.0% (by HPLC)
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Formula: C₁₃H₁₀BrCl₂FN₂O
MDL Number: MFCD18207061
CAS Number: 877399-00-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Specific rotation (value) -64.0 to -70.0 deg (C=1, CHCL3)
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 108.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR