Dye Pipettes, Glass, Color-Coded
Supplier: DWK Life Sciences
PF8144K PF8144M PF8144F PF8144N
53383-336PK 684.21 CAD
53383-336 53383-330 53383-332 53383-334
Dye Pipettes, Glass, Color-Coded
Pipettes Serological Pipettes
Calibrated to deliver with blowout.
Made of tough borosilicate glass. Color-coded for quick, accurate identification.
Perfect for pipetting dark color dye solutions. Pipettes feature an inverse scale with volume graduations starting at zero at tip and are easier to read than conventional amber markings.