5'-Iodo-m-terphenyl ≥98.0% (by GC)

Supplier: TCI America

I0969-1G I0969-200MG
TCI0969-1GEA 545.42 CAD
TCI0969-1G TCI0969-200MG
5'-Iodo-m-terphenyl ≥98.0% (by GC)
5'-Iodo-m-terphenyl
Formula: C₁₈H₁₃I
MDL Number: MFCD26131291
CAS Number: 87666-86-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR