Coumarin 510 ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,3,6,7-Tetrahydro-10-(3-pyridyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one

C3072-1G C3072-200MG
TCC3072-1GEA 546.68 CAD
TCC3072-1G TCC3072-200MG
Coumarin 510 ≥98.0% (by HPLC, total nitrogen)
Coumarin 510
Formula: C₂₀H₁₈N₂O₂
MW: 318.38 g/mol
MDL Number: MFCD00467042
CAS Number: 87349-92-6

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Molar extinction coefficient min. 34000 (THF, 412 to 417nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR