4-(1-Pyrenyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
P2305-1G
P2305-5G
TCP2305-1GEA
265.44
CAD
TCP2305-1G
TCP2305-5G
4-(1-Pyrenyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Pyrenyl)phenylboronic acid
Formula:
C₂₂H₁₅BO₂ |
MDL Number:
MFCD28130386 CAS Number: 872050-52-7 |
Specification Test Results
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (Neutralization titration) | 95.0 to 105.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...