9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 9-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole, 2-[3-(9H-Carbazol-9-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(9H-Carbazol-9-yl)phenylboronic acid Pinacol ester

T3386-1G T3386-5G
TCT3386-1GEA 145.36 CAD
TCT3386-1G TCT3386-5G
9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole ≥98.0% (by GC)
9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
Formula: C₂₄H₂₄BNO₂
MW: 369.27 g/mol
MDL Number: MFCD22415352
CAS Number: 870119-58-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR