9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-Bromo-10-(4-(naphthalen-2-yl)phenyl)anthracene

B4571-1G B4571-200MG
TCB4571-1GEA 528.98 CAD
TCB4571-1G TCB4571-200MG
9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
9-Bromo-10-[4-(2-naphthyl)phenyl]anthracene
Formula: C₃₀H₁₉Br
MW: 459.39 g/mol
MDL Number: MFCD20486476
CAS Number: 866611-29-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR