4-(3-Bromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B4930-1G B4930-200MG
TCB4930-1GEA 109.02 CAD
TCB4930-1G TCB4930-200MG
4-(3-Bromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
4-(3-Bromophenyl)-2,6-diphenylpyrimidine
Formula: C₂₂H₁₅BrN₂
MDL Number: MFCD28962150
CAS Number: 864377-28-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR