Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

P2410-1G P2410-5G
TCP2410-1GEA 200.66 CAD
TCP2410-1G TCP2410-5G
Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide ≥97.0% (by titrimetric analysis)
Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide
Formula: CF₄KNO₄S₂
Melting Pt: 101°C
MDL Number: MFCD28334505
CAS Number: 860653-59-4
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR