2-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5611-1G
B5611-5G
TCB5611-1GEA
246.48
CAD
TCB5611-1G
TCB5611-5G
2-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)
2-Bromo-9-(2-ethylhexyl)-9H-carbazole
Formula:
C₂₀H₂₄BrN MW: 358.32 g/mol |
CAS Number:
856422-39-4 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...