Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate monohydrate ≥98.0% (by GC)

Supplier: TCI America

D4925-1G D4925-5G
TCD4925-1GEA 271.76 CAD
TCD4925-1G TCD4925-5G
Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate monohydrate ≥98.0% (by GC)
Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate monohydrate
Formula: C₉H₉NO₅·H₂O
MDL Number: MFCD00185834
CAS Number: 855740-52-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Water 6.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR