4-(alpha-Methylbenzyl)resorcinol ≥98.0% (by GC)

Supplier: TCI America
Warning

M2754-1G M2754-5G
TCM2754-1GEA 88.4 CAD
TCM2754-1G TCM2754-5G
4-(alpha-Methylbenzyl)resorcinol ≥98.0% (by GC)
4-(alpha-Methylbenzyl)resorcinol
Formula: C₁₄H₁₄O₂
Boiling Pt: 244°C
MDL Number: MFCD21648466
CAS Number: 85-27-8
EINECS: 480-070-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR