1-Chloro-2,4-bis(hexyloxy)benzene ≥96.0% (by GC)

Supplier: TCI America

C3206-1G C3206-5G
TCC3206-1GEA 153.73 CAD
TCC3206-1G TCC3206-5G
1-Chloro-2,4-bis(hexyloxy)benzene ≥96.0% (by GC)
1-Chloro-2,4-bis(hexyloxy)benzene
Formula: C₁₈H₂₉ClO₂
MDL Number: MFCD29089351
CAS Number: 851228-26-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR