1-Methyl-3-propylimidazolium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M3034-25G M3034-5G
76220-326EA 320.74 CAD
76220-326 76220-328
1-Methyl-3-propylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium bromide
Formula: C₇H₁₃BrN₂
MDL Number: MFCD17014727
CAS Number: 85100-76-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR