Altrenogest ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Allyltrenbolone, 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one

A2892-1G A2892-200MG
TCA2892-1GEA 306.52 CAD
TCA2892-1G TCA2892-200MG
Altrenogest ≥98.0% (by HPLC)
Altrenogest
Formula: C₂₁H₂₆O₂
MW: 310.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00867855
CAS Number: 850-52-2
EINECS: 212-703-1
Merck Index: 14,00319

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D -72.0 to -77.0 deg (C=0.5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR