N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-[Bis(4-bromophenyl)amino]phenylboronic acid Pinacol ester, 2-[4-[Bis(4-bromophenyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Bromo-N-(4-bromophenyl)-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)aniline

B4660-1G B4660-200MG
TCB4660-1GEA 884.8 CAD
TCB4660-1G TCB4660-200MG
N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formula: C₂₄H₂₄BBr₂NO₂
MW: 529.08 g/mol
MDL Number: MFCD30721938
CAS Number: 850153-24-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR