(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D3843-1G D3843-5G
76219-690EA 323.16 CAD
76219-690 76219-692
(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)
(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether
Formula: C₂₀H₂₇NOSi
Storage Temperature: Refrigerator
MDL Number: MFCD08690083
CAS Number: 848821-58-9

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -48.0 to -53.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR