2-Bromophenyl p-Toluenesulfonate ≥98.0% (by GC)

Supplier: TCI America

B4528-1G B4528-5G
TCB4528-1GEA 77.74 CAD
TCB4528-1G TCB4528-5G
2-Bromophenyl p-Toluenesulfonate ≥98.0% (by GC)
2-Bromophenyl p-Toluenesulfonate
Formula: C₁₃H₁₁BrO₃S
MDL Number: MFCD19707445
CAS Number: 84672-48-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR