trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

P2404-1G P2404-5G
TCP2404-1GEA 50.56 CAD
TCP2404-1G TCP2404-5G
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl
Formula: C₂₂H₃₄
MDL Number: MFCD13182311
CAS Number: 84656-75-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 107 to 182 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR