4-(6-Hydroxyhexyloxy)benzoic acid ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-((6-Hydroxyhexyl)oxy)benzoic acid

H1629-1G H1629-5G
TCH1629-1GEA 132.72 CAD
TCH1629-1G TCH1629-5G
4-(6-Hydroxyhexyloxy)benzoic acid ≥97.0% (by GC)
4-(6-Hydroxyhexyloxy)benzoic acid
Formula: C₁₃H₁₈O₄
MW: 238.28 g/mol
MDL Number: MFCD00971624
CAS Number: 83883-25-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR