Sultamicillin Tosylate dihydrate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
U0097-1G
U0097-250MG
TCU0097-1GEA
398.16
CAD
TCU0097-1G
TCU0097-250MG
Sultamicillin Tosylate dihydrate ≥98.0% (by GC, titration analysis)
Sultamicillin Tosylate dihydrate
Formula:
C₂₅H₃₀N₄O₉S₂·C₇H₈O₃S·2H₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD01682151 CAS Number: 83105-70-8 Merck Index: 14,08992 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +168 to +186 deg (C=0.5, water/CH3CN 3:2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...