N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N2,N4,N6-tri-m-tolyl-1,3,5-Triazine-2,4,6-triamine, Tris(3-methylanilino)triazine

T3240-1G T3240-200MG
TCT3240-1GEA 350.76 CAD
TCT3240-1G TCT3240-200MG
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine ≥98.0% (by HPLC, total nitrogen)
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine
Formula: C₂₄H₂₄N₆
MW: 396.50 g/mol
Melting Pt: 233°C
MDL Number: MFCD19440763
CAS Number: 82504-70-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR