9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
O0404-25G
O0404-5G
TCO0404-25GEA
431.34
CAD
TCO0404-25G
TCO0404-5G
9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid ≥98.0% (by HPLC)
9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
Formula:
C₁₃H₉F₂NO₄ Melting Pt: 311°C |
MDL Number:
MFCD00226106 CAS Number: 82419-35-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...