3-Hydroxy-4,4'-dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
H1556-1G
H1556-200MG
TCH1556-1GEA
1318.98
CAD
TCH1556-1G
TCH1556-200MG
3-Hydroxy-4,4'-dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
3-Hydroxy-4,4'-dimethyl-2,2'-bipyridyl
Formula:
C₁₂H₁₂N₂O |
MDL Number:
MFCD28339195 CAS Number: 81998-07-4 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...