1,4,5,8-Tetrachloroanthraquinone ≥95.0% (by GC)

Supplier: TCI America
Warning

T3300-25G T3300-5G
76220-554EA 513.5 CAD
76220-554 76220-556
1,4,5,8-Tetrachloroanthraquinone ≥95.0% (by GC)
1,4,5,8-Tetrachloroanthraquinone
Formula: C₁₄H₄Cl₄O₂
Melting Pt: 342°C
MDL Number: MFCD00035801
CAS Number: 81-58-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR