7-Amino-2-benzoxazolinone ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2806-1G A2806-250MG
TCA2806-1GEA 181.86 CAD
TCA2806-1G TCA2806-250MG
7-Amino-2-benzoxazolinone ≥95.0% (by GC, titration analysis)
7-Amino-2-benzoxazolinone
Formula: C₇H₆N₂O₂
MDL Number: MFCD09753616
CAS Number: 81282-60-2
EINECS: 462-910-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR