4-(tert-Butyl)-2-chloropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5466-1G B5466-5G
TCB5466-1GEA 240.16 CAD
TCB5466-1G TCB5466-5G
4-(tert-Butyl)-2-chloropyridine ≥98.0% (by GC)
4-(tert-Butyl)-2-chloropyridine
Formula: C₉H₁₂ClN
MDL Number: MFCD12131129
CAS Number: 81167-60-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR