2-Bromo-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4939-1G B4939-5G
TCB4939-1GEA 197.5 CAD
TCB4939-1G TCB4939-5G
2-Bromo-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
2-Bromo-4,6-diphenyl-1,3,5-triazine
Formula: C₁₅H₁₀BrN₃
MW: 312.17 g/mol
MDL Number: MFCD10687147
CAS Number: 80984-79-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR