N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-Succinimidyl (Boc-aminooxy)acetate, 2,5-Dioxopyrrolidin-1-yl 2-(((tert-butoxycarbonyl)amino)oxy)acetate, (Boc-aminooxy)acetic acid N-Succinimidyl ester, [(tert-Butoxycarbonyl)aminooxy]acetic acid N-Succinimidyl ester, Boc-NH-O-C1-NHS ester

U0096-1G U0096-200MG
TCU0096-1GEA 570.38 CAD
TCU0096-1G TCU0096-200MG
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate ≥98.0% (by HPLC, total nitrogen)
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate
Formula: C₁₁H₁₆N₂O₇
MW: 288.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18382162
CAS Number: 80366-85-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR