2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) ≥98.0% (by GC)

Supplier: TCI America
Warning

O0469-5G O0469-25G
76220-186EA 320.74 CAD
76220-186 76220-394
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) ≥98.0% (by GC)
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane)
Formula: C₁₀H₁₀F₈O₂
Boiling Pt: 112°C
Flash Pt: 133°C
MDL Number: MFCD01320774
CAS Number: 791-22-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR