4-Bromo-2-(trimethylsilyl)thiophene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4856-1G B4856-200MG
TCB4856-1GEA 635.16 CAD
TCB4856-1G TCB4856-200MG
4-Bromo-2-(trimethylsilyl)thiophene ≥97.0% (by GC)
4-Bromo-2-(trimethylsilyl)thiophene
Formula: C₇H₁₁BrSSi
Storage Temperature: Refrigerator
CAS Number: 77998-61-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR