1-Methyl-2-(2-pyridyl)pyridinium iodide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2778-1G M2778-200MG
76220-254EA 627.26 CAD
76220-254 76220-256
1-Methyl-2-(2-pyridyl)pyridinium iodide ≥98.0% (by HPLC, titration analysis)
1-Methyl-2-(2-pyridyl)pyridinium iodide
Formula: C₁₁H₁₁IN₂
MDL Number: MFCD23700876
CAS Number: 77972-47-5

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR